Learn More
Biolegend Brilliant Violet 711™ anti-human CD159a (NKG2A) [S19004C]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 100 tests
Catalog No. 502723228
Encompass_Preferred
Brilliant Violet 711™ anti-human CD159a (NKG2A) [S19004C]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 100 tests
Catalog No. 50-272-3228 Supplier Biolegend Supplier No. 375156
Brilliant Violet 711™ anti-human CD159a (NKG2A) [S19004C]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 100 tests