Learn More
Biolegend Brilliant Violet 711™ anti-human CD135 (Flt-3/Flk-2) [BV10A4H2]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 100 tests
Supplier:  Biolegend 313328
Encompass_Preferred
Brilliant Violet 711™ anti-human CD135 (Flt-3/Flk-2) [BV10A4H2]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 100 tests
Catalog No. 50-272-3349