Learn More
Biolegend Brilliant Violet 711TM anti-hu
Supplier:  Biolegend 386811
Encompass_Preferred
Brilliant Violet 711™ anti-human CD66b [W20104A]; Isotype: Rat IgG2a, κ; Reactivity: Human; Apps: FC; Size: 25 tests
Catalog No. 50-274-5578