Learn More
Biolegend Brilliant Violet 711TM anti-hu
Supplier:  Biolegend 330939
Encompass_Preferred
Brilliant Violet 711 anti-human CD58 (LFA-3) [TS2/9] Isotype Mouse IgG1 Reactivity Human Apps FC Size 25 tests
Catalog No. 50-003-16020