Learn More
Biolegend Brilliant Violet 711TM anti-mo
Catalog No. 502722812
Encompass_Preferred
Brilliant Violet 711™ anti-mouse CD24 [M1/69]; Isotype: Rat IgG2b, κ; Reactivity: Mouse; Apps: FC; Size: 50 μg
Catalog No. 50-272-2812 Supplier Biolegend Supplier No. 101851
Brilliant Violet 711™ anti-mouse CD24 [M1/69]; Isotype: Rat IgG2b, κ; Reactivity: Mouse; Apps: FC; Size: 50 μg