Learn More
Biolegend Brilliant Violet 750TM anti-mo
Supplier:  Biolegend 103079
Encompass_Preferred
Brilliant Violet 750™ anti-mouse/human CD44 [IM7]; Isotype: Rat IgG2b, κ; Reactivity: Mouse, Human; Apps: FC; Size: 50 μg
Catalog No. 50-272-2804