Learn More
Biolegend Brilliant Violet 785™ anti-human CD25
Supplier:  Biolegend 356140
Encompass_Preferred
Brilliant Violet 785™ anti-human CD25 [M-A251]; Isotype: Mouse IgG1, κ; Reactivity: Human, Cross-Reactivity: Baboon, Cynomolgus, Rhesus; Apps: FC; Size: 100 tests
Catalog No. NC1569611