Learn More
Biolegend Brilliant Violet 785™ anti-human CD83
Supplier:  Biolegend 305338
Encompass_Preferred
Brilliant Violet 785™ anti-human CD83 [HB15e]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon, Pigtailed Macaque, Rhesus; Apps: FC; Size: 100 tests
Catalog No. NC1559834