Learn More
Biolegend Brilliant Violet 785™ anti-human CD337 (NKp30)
Supplier:  Biolegend 325230
Encompass_Preferred
Brilliant Violet 785™ anti-human CD337 (NKp30) [P30-15]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon, Cynomolgus, Rhesus; Apps: FC; Size: 100 tests
Catalog No. NC1691355