Learn More
Biolegend Brilliant Violet 785™ anti-mouse CD4
Supplier:  Biolegend 100453
Encompass_Preferred
Brilliant Violet 785™ anti-mouse CD4 [GK1.5]; Isotype: Rat IgG2b, κ; Reactivity: Mouse; Apps: FC; Size: 50 ug
Catalog No. NC1027792