Learn More
Biolegend Brilliant Violet 785™ anti-T-bet
Supplier:  Biolegend 644835
Encompass_Preferred
Brilliant Violet 785™ anti-T-bet [4B10]; Isotype: Mouse IgG1, κ; Reactivity: Human, Mouse; Apps: ICFC; Size: 50 ug
Catalog No. NC1543771