Learn More
Biolegend Brilliant Violet 785™ anti-human EGFR [AY13]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 25 tests
Catalog No. 502723272
Encompass_Preferred
Brilliant Violet 785™ anti-human EGFR [AY13]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 25 tests
Catalog No. 50-272-3272 Supplier Biolegend Supplier No. 352941
Brilliant Violet 785™ anti-human EGFR [AY13]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 25 tests