Learn More
Biolegend Brilliant Violet 785TM anti-mo
Catalog No. 502721887
Encompass_Preferred
Brilliant Violet 785™ anti-mouse IFN-γ [XMG1.2]; Isotype: Rat IgG1, κ; Reactivity: Mouse; Apps: ICFC; Size: 125 μL
Catalog No. 50-272-1887 Supplier Biolegend Supplier No. 505837
Brilliant Violet 785™ anti-mouse IFN-γ [XMG1.2]; Isotype: Rat IgG1, κ; Reactivity: Mouse; Apps: ICFC; Size: 125 μL