Learn More
Jackson Immuno Research Labs Brilliant Violet 421™ AffiniPure Donkey Anti-Rabbit IgG (H+L), 0.25 ml
SDP
Catalog No. NC1687676
Encompass_Preferred
Whole IgG antibodies are isolated as intact molecules from antisera by immunoaffinity chromatography. They have an Fc portion and two antigen binding Fab portions joined together by disulfide bonds and therefore they are divalent. The average molecular weight is reported to be about 160 kDa. The whole IgG form of antibodies is suitable for the majority of immunodetection procedures and is the most cost effective.
Catalog No. NC1687676 Supplier Jackson Immuno Research Labs Supplier No. 711675152
Whole IgG antibodies are isolated as intact molecules from antisera by immunoaffinity chromatography. They have an Fc portion and two antigen binding Fab portions joined together by disulfide bonds and therefore they are divalent. The average molecular weight is reported to be about 160 kDa. The whole IgG form of antibodies is suitable for the majority of immunodetection procedures and is the most cost effective.