Learn More
Biolegend Brilliant Violet 711™ anti-human CD33
Supplier:  Biolegend 303424
Encompass_Preferred
Brilliant Violet 711™ anti-human CD33 [WM53]; Isotype: Mouse IgG1, κ; Reactivity: Human, Chimpanzee; Apps: FC; Size: 100 tests
Catalog No. NC1043860