Learn More
Medchemexpress LLC Brilliant Yellow 25g
SDP
Catalog No. 5000206150
Encompass_Preferred
Brilliant Yellow a diazo-containing sulfonic aci is also a potent VGLUT-specific inhibitor Brilliant Yellow is membrane-impermeable However there are some Brilliant Yellow analogs with low cytotoxicity and cell penetration Brilliant Yellow analogs work on glutamatergic transmission in hippocampal neurons[1]
Catalog No. 50-002-06150 Supplier Medchemexpress LLC Supplier No. 25GHYW110888
Brilliant Yellow a diazo-containing sulfonic aci is also a potent VGLUT-specific inhibitor Brilliant Yellow is membrane-impermeable However there are some Brilliant Yellow analogs with low cytotoxicity and cell penetration Brilliant Yellow analogs work on glutamatergic transmission in hippocampal neurons[1]