Learn More
Sigma Aldrich Fine Chemicals Biosciences Brilliant Yellow Dye conte25G
Catalog No. 502900013
Encompass
Brilliant Yellow is an orange colored powder and it belongs to the group of azo dyes. It is yellow at pH 6.4 and red-orange at pH 8.0. Brilliant Yellow has been widely used as a biosensor anti-fungal agent anti-AIDS (acquired immunodeficiency syndrome) agent and is used in assays for enzyme activity.
Catalog No. 50-290-0013 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 20137525G
Brilliant Yellow is an orange colored powder and it belongs to the group of azo dyes. It is yellow at pH 6.4 and red-orange at pH 8.0. Brilliant Yellow has been widely used as a biosensor anti-fungal agent anti-AIDS (acquired immunodeficiency syndrome) agent and is used in assays for enzyme activity.