Learn More
General Laboratory Products Brilliant Green Bile Broth (BGBB) 2% w/ Durham - 10ml, 20/pk
Catalog No. NC1689469
Encompass_Preferred
2% Brilliant Green Bile Broth w/ Durham tube - 10ml, 16x150mm glass tube, 20 per pack. Base for Confirming the Presence of Coliform Organisms in Water and Foods.
Catalog No. NC1689469 Supplier General Laboratory Products Supplier No. 16GL11001020PK
2% Brilliant Green Bile Broth w/ Durham tube - 10ml, 16x150mm glass tube, 20 per pack. Base for Confirming the Presence of Coliform Organisms in Water and Foods.