Learn More
Biolegend Brilliant Violet 421™ anti-human CD3
Supplier:  Biolegend 317343
Encompass_Preferred
Brilliant Violet 421™ anti-human CD3 [OKT3]; Isotype: Mouse IgG2a, κ; Reactivity: Human; Apps: FC; Size: 25 tests
Catalog No. NC0923380