Learn More
Biolegend Brilliant Violet 421™ anti-human CD15 (SSEA-1)
Supplier:  Biolegend 323039
Encompass_Preferred
Brilliant Violet 421™ anti-human CD15 (SSEA-1) [W6D3]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 25 tests
Catalog No. NC1583991