Learn More
Cayman Chemical BrImonIdIn-d4 1mg
SDP
Supplier:  Cayman Chemical 254651 MG
Encompass_Preferred
An internal standard for the quantification of brimonidine by GC- or LC-MS
Catalog No. 50-293-6037