Learn More
Cayman Chemical BrImonIdIn-d4 5mg
SDP
Supplier:  Cayman Chemical 254655 MG
Encompass_Preferred
An internal standard for the quantification of brimonidine by GC- or LC-MS
Catalog No. 50-293-6573