Learn More
Medchemexpress LLC HY-B0659 10mg Medchemexpress, Brimonidine CAS:59803-98-4 Purity:>98%
SDP
Catalog No. 502082634
Encompass_Preferred
Medchemexpress, HY-B0659 10mg Brimonidine CAS:59803-98-4 Brimonidine (UK 14304) is a full α2-adrenergic receptor (α2-AR) agonist. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2634 Supplier Medchemexpress LLC Supplier No. HYB065910MG
Medchemexpress, HY-B0659 10mg Brimonidine CAS:59803-98-4 Brimonidine (UK 14304) is a full α2-adrenergic receptor (α2-AR) agonist. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.