Learn More
Medchemexpress LLC Brimonidine (Synonyms: UK 14304; AGN190342) Solution 10 mM * 1 mL in DMSO
SDP
Catalog No. NC3781236
Encompass_Preferred
Brimonidine (UK 14304) is a full α2-adrenergic receptor (α2-AR) agonist.
Catalog No. NC3781236 Supplier Medchemexpress LLC Supplier No. HYB065910MM 1ML/SOLUTN
Brimonidine (UK 14304) is a full α2-adrenergic receptor (α2-AR) agonist.