Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Brimonidine, 98%

Catalog No. AC461520010
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC461520010 1 g
AC461522500 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC461520010 Supplier Thermo Scientific Chemicals Supplier No. 461520010
Add to Cart

CAS: 59803-98-4 | C11H10BrN5 | 292.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 59803-98-4
Molecular Formula C11H10BrN5
Molecular Weight (g/mol) 292.14
MDL Number MFCD00153878
InChI Key XYLJNLCSTIOKRM-UHFFFAOYSA-N
Synonym brimonidine, bromoxidine, mirvaso, 5-bromo-n-4,5-dihydro-1h-imidazol-2-yl-6-quinoxalinamine, 5-bromo-n-4,5-dihydro-1h-imidazol-2-yl quinoxalin-6-amine, 5-bromo-6-2-imidazolin-2-ylamino quinoxaline, unii-e6gnx3hhte, brimonidine inn:ban, 3h brimonidine, 5-bromo-6-imidazolin-2-ylamino quinoxaline
PubChem CID 2435
ChEBI CHEBI:3175
IUPAC Name 5-bromo-N-(4,5-dihydro-1H-imidazol-2-yl)quinoxalin-6-amine
SMILES BrC1=C2N=CC=NC2=CC=C1NC1=NCCN1

Specifications

Melting Point 235.0°C to 236.0°C
Ignition Residue 0.3% max.
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Solubility Information Solubility in water: insoluble.
Formula Weight 292.13
Percent Purity 98%
Chemical Name or Material Brimonidine
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement Toxic if swallowed.
Precautionary Statement GHS P Statement Wash face, hands and any exposed skin thoroughly after handling. IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.