Learn More
Apexbio Technology LLC Brimonidine Tartrate 70359-46-5 50mg
SDP
Catalog No. 502240799
Encompass_Preferred
Brimonidine tartrate (CAS 70359-46-5) is a selective agonist of the -adrenergic receptor By activating these receptors it inhibits adenylyl cyclase activity resulting in decreased intracellular cyclic AMP (cAMP) levels This modulation reduces sympathetic neural activity lowers aqueous humor production and enhances uveoscleral outflow in ocular tissues Brimonidine tartrate is widely employed in research on glaucoma and ocular hypertension and serves as a standard tool in studies investigating -adrenergic signaling and autonomic nervous system regulation
This item is not returnable. View return policy
Catalog No. 50-224-0799 Supplier Apexbio Technology LLC Supplier No. B168350
Brimonidine tartrate (CAS 70359-46-5) is a selective agonist of the -adrenergic receptor By activating these receptors it inhibits adenylyl cyclase activity resulting in decreased intracellular cyclic AMP (cAMP) levels This modulation reduces sympathetic neural activity lowers aqueous humor production and enhances uveoscleral outflow in ocular tissues Brimonidine tartrate is widely employed in research on glaucoma and ocular hypertension and serves as a standard tool in studies investigating -adrenergic signaling and autonomic nervous system regulation