Learn More
Selleck Chemical LLC Brimonidine Tartrate S4266-1g
SDP
Catalog No. 503109455
Encompass_Preferred
Brimonidine Tartrate(UK 1434 Tartrate) is a highly selective -adrenergic receptor agonist with EC50 of 0 45 nM for the 2A adrenoreceptor and used to treat open-angle glaucoma or ocular hypertension
Catalog No. 50-310-9455 Supplier Selleck Chemical LLC Supplier No. S42661G
Brimonidine Tartrate(UK 1434 Tartrate) is a highly selective -adrenergic receptor agonist with EC50 of 0 45 nM for the 2A adrenoreceptor and used to treat open-angle glaucoma or ocular hypertension