Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brimonidine tartrate, Thermo Scientific™

Catalog No. AC468842500
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC468842500 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AC468842500 Supplier Thermo Scientific Chemicals Supplier No. 468842500
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 70359-46-5
Molecular Formula C15H16BrN5O6
Molecular Weight (g/mol) 442.23
MDL Number 07773072
InChI Key QZHBYNSSDLTCRG-LREBCSMRSA-N
IUPAC Name (2R,3R)-2,3-dihydroxybutanedioic acid; 5-bromo-N-(4,5-dihydro-1H-imidazol-2-yl)quinoxalin-6-amine
SMILES O[C@H]([C@@H](O)C(O)=O)C(O)=O.BrC1=C2N=CC=NC2=CC=C1NC1=NCCN1

Specifications

Packaging Glass Bottle
Certificate of Analysis Available
Physical Form Powder
Color White to Light Yellow to Light Brown
Melting Point 207°C
Solubility Soluble in water
Quantity 250 mg
Chemical Name or Material Brimonidine tartrate
Hazard Category Skull and crossbones
Recommended Storage Normal conditions
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.