Learn More
Cayman Chemical Brimonidine (tartrate), Purity: ≥98%, CAS: 70359-46-5, 100 mg
SDP
Catalog No. 502969122
Encompass_Preferred
An α2-AR agonist (Kis = 2.7, 52, and 44 nM for α2A, α2B, and α2C-ARs, respectively, in CHO cells); selective for α2-ARs over α1-ARs (Ki = 1,800 nM in human brain); lowers intraocular pressure in DBA/2J mice to control levels at 0.1% applied topically to the eye; protects rat retinal ganglion cells against glutamate excitotoxicity in a rat model of retinal ischemia at 1 mg/kg per day
Catalog No. 50-296-9122 Supplier Cayman Chemical Supplier No. 15426100 MG
An α2-AR agonist (Kis = 2.7, 52, and 44 nM for α2A, α2B, and α2C-ARs, respectively, in CHO cells); selective for α2-ARs over α1-ARs (Ki = 1,800 nM in human brain); lowers intraocular pressure in DBA/2J mice to control levels at 0.1% applied topically to the eye; protects rat retinal ganglion cells against glutamate excitotoxicity in a rat model of retinal ischemia at 1 mg/kg per day