Learn More
Cayman Chemical Brincidofovir, Purity: ≥98%, CAS: 444805-28-1, 50 mg
SDP
Catalog No. 502911500
Encompass_Preferred
A prodrug form of cidofovir; inhibits CMV, HSV-1, HSV-2, cowpox, and vaccinia virus replication in plaque reduction assays (EC50s = 0.0009, 0.06, 0.08, 0.6, and ~1 µM, respectively); decreases mortality in mouse models of cowpox or vaccinia virus infections at 6.7 or 5 mg/kg, respectively
Catalog No. 50-291-1500 Supplier Cayman Chemical Supplier No. 3506250 MG
A prodrug form of cidofovir; inhibits CMV, HSV-1, HSV-2, cowpox, and vaccinia virus replication in plaque reduction assays (EC50s = 0.0009, 0.06, 0.08, 0.6, and ~1 µM, respectively); decreases mortality in mouse models of cowpox or vaccinia virus infections at 6.7 or 5 mg/kg, respectively