Learn More
Selleck Chemical LLC Brincidofovir-E0938-5MG
SDP
Catalog No. 502389985
Encompass_Preferred
Brincidofovir (CMX001) the lipid-conjugated prodrug of Cidofovir with long-acting antiviral effect shows activity against a broad spectrum of DNA viruses including cytomegalovirus (CMV) adenovirus (ADV) varicella zoster virus herpes simplex virus polyomaviruses papillomaviruses poxviruses and mixed double-stranded DNA virus infections
Catalog No. 50-238-9985 Supplier Selleck Chemical LLC Supplier No. E09385MG
Brincidofovir (CMX001) the lipid-conjugated prodrug of Cidofovir with long-acting antiviral effect shows activity against a broad spectrum of DNA viruses including cytomegalovirus (CMV) adenovirus (ADV) varicella zoster virus herpes simplex virus polyomaviruses papillomaviruses poxviruses and mixed double-stranded DNA virus infections