Learn More
AdipoGen Brinzolamide
SDP
Numéro de catalogue. 502561595
Encompass_Preferred
Chemical. CAS 138890-62-7. Formula C12H21N3O5S3. MW 383.51. Synthetic. Brinzolamide is a potent inhibitor of carbonic anhydrases IC50=3.2nM and 45.3nM for CAII and CAIV, respectively that lowers intraocular pressure by locally inhibiting carbonic anhydrase in the ciliary processes and suppressing aqueous humor secretion. Animal studies have found that one week of twice-daily topical ocular treatment with 2% brinzolamide significantly reduced IOP and enhanced optic nerve head blood flow.
Numéro de catalogue. 50-256-1595 Fournisseur AdipoGen Code fournisseur CDXB0424M010
Ajouter au panier
Ajouter au panier
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
Chemical. CAS 138890-62-7. Formula C12H21N3O5S3. MW 383.51. Synthetic. Brinzolamide is a potent inhibitor of carbonic anhydrases IC50=3.2nM and 45.3nM for CAII and CAIV, respectively that lowers intraocular pressure by locally inhibiting carbonic anhydrase in the ciliary processes and suppressing aqueous humor secretion. Animal studies have found that one week of twice-daily topical ocular treatment with 2% brinzolamide significantly reduced IOP and enhanced optic nerve head blood flow.
missing translation for 'productTitle'
missing translation for 'pleaseSelectIssue'
missing translation for 'byClickingSubmit' missing translation for 'privacyPolicy'.