Learn More
Apexbio Technology LLC Brinzolamide 138890-62-7 10mM (in 1mL DMSO)
SDP
Catalog No. 5000094781
Encompass_Preferred
Brinzolamide (CAS 138890-62-7) is a potent and selective inhibitor of carbonic anhydrase II (CA II) with an IC50 of 3 19 nM and a Ki of 0 13 nM in vitro By inhibiting CA II brinzolamide disrupts the reversible hydration of carbon dioxide thereby reducing aqueous humor production and lowering intraocular pressure Demonstrated to have higher affinity and inhibitory efficacy for CA II compared to CA I and CA IV brinzolamide has been shown to significantly decrease intraocular pressure in animal models Additionally it is utilized in research settings to assess cerebral blood flow reserve as CA inhibition modulates tissue CO2 levels and consequent vasodilation
Catalog No. 50-000-94781 Supplier Apexbio Technology LLC Supplier No. A43595.1
Brinzolamide (CAS 138890-62-7) is a potent and selective inhibitor of carbonic anhydrase II (CA II) with an IC50 of 3 19 nM and a Ki of 0 13 nM in vitro By inhibiting CA II brinzolamide disrupts the reversible hydration of carbon dioxide thereby reducing aqueous humor production and lowering intraocular pressure Demonstrated to have higher affinity and inhibitory efficacy for CA II compared to CA I and CA IV brinzolamide has been shown to significantly decrease intraocular pressure in animal models Additionally it is utilized in research settings to assess cerebral blood flow reserve as CA inhibition modulates tissue CO2 levels and consequent vasodilation