Learn More
Apexbio Technology LLC Brinzolamide 138890-62-7 50mg
SDP
Catalog No. 5000096119
Encompass_Preferred
Brinzolamide (CAS 138890-62-7) is a potent and selective inhibitor of carbonic anhydrase II (CA II) with an IC50 of 3 19 nM and a Ki of 0 13 nM in vitro By inhibiting CA II brinzolamide disrupts the reversible hydration of carbon dioxide thereby reducing aqueous humor production and lowering intraocular pressure Demonstrated to have higher affinity and inhibitory efficacy for CA II compared to CA I and CA IV brinzolamide has been shown to significantly decrease intraocular pressure in animal models Additionally it is utilized in research settings to assess cerebral blood flow reserve as CA inhibition modulates tissue CO2 levels and consequent vasodilation
Catalog No. 50-000-96119 Supplier Apexbio Technology LLC Supplier No. A435950
Brinzolamide (CAS 138890-62-7) is a potent and selective inhibitor of carbonic anhydrase II (CA II) with an IC50 of 3 19 nM and a Ki of 0 13 nM in vitro By inhibiting CA II brinzolamide disrupts the reversible hydration of carbon dioxide thereby reducing aqueous humor production and lowering intraocular pressure Demonstrated to have higher affinity and inhibitory efficacy for CA II compared to CA I and CA IV brinzolamide has been shown to significantly decrease intraocular pressure in animal models Additionally it is utilized in research settings to assess cerebral blood flow reserve as CA inhibition modulates tissue CO2 levels and consequent vasodilation