Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Brinzolamide | 138890-62-7 | 99.9% | 100 MG
SDP

Catalog No. 5000408337
Encompass_Preferred

Brinzolamide (AL-4862) is a selective carbonic anhydrase II inhibitor with an IC50 value of 3.2 nM. It reduces intraocular pressure (IOP) by inhibiting ciliary CA-II and decreasing atrial fluid secretion, and can be used in glaucoma disease research.

  • Selective carbonic anhydrase II inhibitor
  • IC50 value of 3.2 nM
  • Reduces intraocular pressure (IOP)
  • Suitable for glaucoma disease research
  • Molecular weight: 383.51
  • Molecular formula: C12H21N3O5S3
  • Appears as a solid, light yellow to yellow

Catalog No. 50-004-08337 Supplier Medchemexpress LLC Supplier No. HYB0588100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Brinzolamide (AL-4862) is a selective carbonic anhydrase II inhibitor with an IC50 value of 3.2 nM. It reduces intraocular pressure (IOP) by inhibiting ciliary CA-II and decreasing atrial fluid secretion, and can be used in glaucoma disease research.

  • Selective carbonic anhydrase II inhibitor
  • IC50 value of 3.2 nM
  • Reduces intraocular pressure (IOP)
  • Suitable for glaucoma disease research
  • Molecular weight: 383.51
  • Molecular formula: C12H21N3O5S3
  • Appears as a solid, light yellow to yellow

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.