Learn More
Medchemexpress LLC Brinzolamide | 138890-62-7 | 99.9% | 25 MG
SDP
Catalog No. 5000450824
Encompass_Preferred
Brinzolamide (AL-4862) is a selective carbonic anhydrase II inhibitor, demonstrating an IC50 value of 3.2 nM. It effectively reduces intraocular pressure by inhibiting ciliary CA-II and decreasing atrial fluid secretion. This compound is valuable for research in glaucoma disease.
  • Selective carbonic anhydrase II inhibitor
  • IC50 value of 3.2 nM
  • Reduces intraocular pressure (IOP)
  • Useful in glaucoma disease research
Catalog No. 50-004-50824 Supplier Medchemexpress LLC Supplier No. 25MGHYB0588
Brinzolamide (AL-4862) is a selective carbonic anhydrase II inhibitor, demonstrating an IC50 value of 3.2 nM. It effectively reduces intraocular pressure by inhibiting ciliary CA-II and decreasing atrial fluid secretion. This compound is valuable for research in glaucoma disease.
  • Selective carbonic anhydrase II inhibitor
  • IC50 value of 3.2 nM
  • Reduces intraocular pressure (IOP)
  • Useful in glaucoma disease research