Learn More
Selleck Chemical LLC Brinzolamide S3178-10mg
SDP
Catalog No. 503109960
Encompass_Preferred
Brinzolamide (AL-4862) is a potent carbonic anhydrase II inhibitor with IC50 of 3 19 nM
Catalog No. 50-310-9960 Supplier Selleck Chemical LLC Supplier No. S317810MG
Brinzolamide (AL-4862) is a potent carbonic anhydrase II inhibitor with IC50 of 3 19 nM