Learn More
PicoStudio Bristlebots
Description
- A Bristlebot is a small robot made by combining a toothbrush head and a vibrating pager motor.
- Vibrations from the motor travel down the bristles and cause the brush to move and spin on flat surfaces.
- Adjust different parts by bending, twisting, and angling the chenille stems into different positions and see how it affects the movement and direction of the Bristlebot.
- Build time: 3 to 10 minutes.
- Suitable for ages 5+.
- Ideal for camps, scout troops, homeschool, libraries, after-school, STEM/STEAM groups, birthday party, and classroom activities.
- Race or battle them against each other.
Specifications
Specifications
| Includes | 25 Toothbrush Heads, 25 Vibrating Motors (1.5 V), 25 Batteries (1.5 V), 50 Pipe Cleaners, 50 Google Eyes, 25 Double Sided Tape |
| Product Type | STEM Kit |
| Kit Size | 25 Pk. |