Learn More
Medchemexpress LLC HY-112589 5mg Medchemexpress, BRITE-338733 CAS:503105-88-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1125895MG
Encompass_Preferred
Medchemexpress, HY-112589 5mg BRITE-338733 CAS:503105-88-2 BRITE-338733 is a RecA ATPase inhibitor, with an IC50 of 4.7 µM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-0821