Learn More
Cayman Chemical BRITE338733 10mg
SDP
Catalog No. 502918017
Encompass_Preferred
A RecA inhibitor (IC50 = 4.7 µM for ATP hydrolysis in a cell-free assay)
Catalog No. 50-291-8017 Supplier Cayman Chemical Supplier No. 3730210 MG
A RecA inhibitor (IC50 = 4.7 µM for ATP hydrolysis in a cell-free assay)