Learn More
Shockwatch Div Mri Britevent Specimen Thermometer
SDP
Catalog No. NC2598448
Encompass_Preferred
Continuous Display Thermometer for Urine Specimen Collection Cup• Easy to Read Digital Event Strip with Adhesive Backing• Instant Response• Conforms to SAMHSA Guidelines• Individual Split Back (Easy to Remove Backing) • 32-39C• 62 / sheet
Catalog No. NC2598448 Supplier Shockwatch Div Mri Supplier No. 4030B
Continuous Display Thermometer for Urine Specimen Collection Cup• Easy to Read Digital Event Strip with Adhesive Backing• Instant Response• Conforms to SAMHSA Guidelines• Individual Split Back (Easy to Remove Backing) • 32-39C• 62 / sheet