Learn More
STEREOGRAPHICS CORPORATION Britevent Specimen Thermometer
Catalog No. NC2598442
Encompass_Preferred
Continuous Display Thermometer for Urine Specimen Collection Cup• Easy to Read Digital Event Strip with Adhesive Backing• Instant Response• Conforms to SAMHSA Guidelines• Individual Split Back (Easy to Remove Backing) • 88-102F• 62 pieces / sheet
Catalog No. NC2598442 Supplier STEREOGRAPHICS CORPORATION Supplier No. 4030
Continuous Display Thermometer for Urine Specimen Collection Cup• Easy to Read Digital Event Strip with Adhesive Backing• Instant Response• Conforms to SAMHSA Guidelines• Individual Split Back (Easy to Remove Backing) • 88-102F• 62 pieces / sheet