Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Britton-Robinson Buffer, pH 1.98, adjusted to 0.1 M Ionic Strength, Ricca Chemical
SDP

Catalog No. R1154200500 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
500 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R1154200500 500 mL
R11542001A 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. R1154200500 Supplier Ricca Chemical Company Supplier No. R1154200500A
Add to Cart
Add to Cart

Specifications

Chemical Name or Material Britton-Robinson Buffer
Color Colorless Liquid
CAS 7732-18-5, 7447-40-7, 7664-38-2, 10043-35-3, 64-19-7, 1310-73-2
CAS Max % 98.55, 0.6, 0.36, 0.23, 0.22, 0.04
Grade Laboratory
Packaging Natural Poly Bottle
pH 1.98
Quantity 500 mL
Sterility Non-sterile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.