Learn More
Medchemexpress LLC Brivanib | 649735-46-6 | MFCD13194684 | >98.0% | 370.38 g/mol | C19H19FN4O3 | 10 MG
SDP
Catalog No. 5000244960
Encompass_Preferred
Brivanib is an ATP-competitive small-molecule inhibitor of vascular endothelial growth factor receptor 2 (VEGFR2) with an IC50 of 25 nM. It displays moderate activity against VEGFR-1 and FGFR-1 and is highly selective versus PDGFR-β. Supplied for research and analytical use, the compound is provided as a powder with specified storage recommendations.
Catalog No. 50-002-44960 Supplier Medchemexpress LLC Supplier No. HY1033710MG
Brivanib is an ATP-competitive small-molecule inhibitor of vascular endothelial growth factor receptor 2 (VEGFR2) with an IC50 of 25 nM. It displays moderate activity against VEGFR-1 and FGFR-1 and is highly selective versus PDGFR-β. Supplied for research and analytical use, the compound is provided as a powder with specified storage recommendations.