Learn More
Medchemexpress LLC Brivanib | 649735-46-6 | MFCD13194684 | 99.5% | 370.38 g/mol | C19H19FN4O3 | 50 MG
SDP
Catalog No. 5000244574
Encompass_Preferred
Brivanib is an ATP-competitive inhibitor of vascular endothelial growth factor receptor 2 (VEGFR2) supplied for research and analytical applications. It inhibits VEGFR2 with an IC50 of approximately 25 nM and shows activity against related kinases.
  • ATP-competitive VEGFR2 inhibitor (IC50 ≈ 25 nM).
  • Moderate activity against VEGFR-1 and FGFR-1; markedly less active versus PDGFR.
  • Supplied as a solid powder with high purity (~99.5%).
  • Suitable for biochemical and cellular research applications.
  • Stable when stored under recommended conditions for powder and solutions.
Catalog No. 50-002-44574 Supplier Medchemexpress LLC Supplier No. HY1033750MG
Brivanib is an ATP-competitive inhibitor of vascular endothelial growth factor receptor 2 (VEGFR2) supplied for research and analytical applications. It inhibits VEGFR2 with an IC50 of approximately 25 nM and shows activity against related kinases.
  • ATP-competitive VEGFR2 inhibitor (IC50 ≈ 25 nM).
  • Moderate activity against VEGFR-1 and FGFR-1; markedly less active versus PDGFR.
  • Supplied as a solid powder with high purity (~99.5%).
  • Suitable for biochemical and cellular research applications.
  • Stable when stored under recommended conditions for powder and solutions.