Learn More
Medchemexpress LLC HY-10336 5mg Medchemexpress, Brivanib (alaninate) CAS:649735-63-7 Purity:>98%
SDP
Catalog No. 501968085
Encompass_Preferred
Medchemexpress, HY-10336 5mg Brivanib (alaninate) CAS:649735-63-7 Brivanib alaninate is an ATP-competitive inhibitor against VEGFR2 with an IC50 of 25 nM; has moderate potency against VEGFR-1 and FGFR-1, but more than 240-fold against PDGFRβ. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8085 Supplier Medchemexpress LLC Supplier No. HY103365MG
Medchemexpress, HY-10336 5mg Brivanib (alaninate) CAS:649735-63-7 Brivanib alaninate is an ATP-competitive inhibitor against VEGFR2 with an IC50 of 25 nM; has moderate potency against VEGFR-1 and FGFR-1, but more than 240-fold against PDGFRβ. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.