Learn More
Selleck Chemical LLC Brivanib (BMS-540215) 5mg 649735-46-6
SDP
Catalog No. 501360901
Encompass_Preferred
Brivanib is an ATP-competitive inhibitor against VEGFR2 with IC50 of 25 nM, moderate potency against VEGFR-1 and FGFR-1, but >240-fold against PDGFR-?. Phase 3. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-0901 Supplier Selleck Chemical LLC Supplier No. S10845MG
Add to Cart
Add to Cart
Brivanib is an ATP-competitive inhibitor against VEGFR2 with IC50 of 25 nM, moderate potency against VEGFR-1 and FGFR-1, but >240-fold against PDGFR-?. Phase 3. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.