Learn More
Cayman Chemical Brivanib Alaninate, Purity: ≥98%, CAS: 649735-63-7, 50 mg
SDP
Catalog No. 502933327
Encompass_Preferred
A prodrug form of brivanib; reduces hepatic angiogenesis and VEGF, TGF-β1, CD31, and phosphorylated VEGFR2 and FGFR levels in a bile duct-ligated rat model of cirrhosis at 3 mg/kg per day; reduces tumor growth in an L2987 non-small cell lung cancer mouse xenograft model, exhibiting 97% tumor growth inhibition at 107 mg/kg
Catalog No. 50-293-3327 Supplier Cayman Chemical Supplier No. 2608150 MG
A prodrug form of brivanib; reduces hepatic angiogenesis and VEGF, TGF-β1, CD31, and phosphorylated VEGFR2 and FGFR levels in a bile duct-ligated rat model of cirrhosis at 3 mg/kg per day; reduces tumor growth in an L2987 non-small cell lung cancer mouse xenograft model, exhibiting 97% tumor growth inhibition at 107 mg/kg