Learn More
Selleck Chemical LLC Brivudine 25mg 69304-47-8 BVDU
SDP
Supplier:  Selleck Chemical LLC S500925MG
Encompass_Preferred
Brivudine (BVDU) is a uridine derivative and nucleoside analog with pro-apoptotic and chemosensitizing properties. It is incorporated into the viral DNA and blocks the action of DNA polymerases, thus inhibiting viral replication. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8398