Learn More
Medchemexpress LLC BRL-15572 dihydrochloride | 193611-72-2 | 99.8% | 479.87 | C25H29Cl3N2O | 5 MG
SDP
Catalog No. 501969836
Encompass_Preferred
BRL-15572 dihydrochloride is a selective antagonist of the human 5-HT1D receptor supplied for research use. The compound is available as a solid in multiple pack sizes and as a 10 mM solution in DMSO, and is characterized by manufacturer-provided analytical data including molecular weight and purity.
Catalog No. 50-196-9836 Supplier Medchemexpress LLC Supplier No. HY132005MG
BRL-15572 dihydrochloride is a selective antagonist of the human 5-HT1D receptor supplied for research use. The compound is available as a solid in multiple pack sizes and as a 10 mM solution in DMSO, and is characterized by manufacturer-provided analytical data including molecular weight and purity.